Merck
  • Vitamin D Signaling Suppresses Early Prostate Carcinogenesis in TgAPT121 Mice.
Vitamin D Signaling Suppresses Early Prostate Carcinogenesis in TgAPT121 Mice.
Cancer prevention research (Philadelphia, Pa.) (2019-04-28)
James C Fleet, Pavlo L Kovalenko, Yan Li, Justin Smolinski, Colleen Spees, Jun-Ge Yu, Jennifer M Thomas-Ahner, Min Cui, Antonio Neme, Carsten Carlberg, Steven K Clinton
ABSTRACT
We tested whether lifelong modification of vitamin D signaling can alter the progression of early prostate carcinogenesis in studies using mice that develop high-grade prostatic intraepithelial neoplasia that is similar to humans. Two tissue-limited models showed that prostate vitamin D receptor (VDR) loss increased prostate carcinogenesis. In another study, we fed diets with three vitamin D3 levels (inadequate = 25 IU/kg diet, adequate for bone health = 150 IU/kg, or high = 1,000 IU/kg) and two calcium levels (adequate for bone health = 0.5% and high = 1.5%). Dietary vitamin D caused a dose-dependent increase in serum 25-hydroxyvitamin D levels and a reduction in the percentage of mice with adenocarcinoma but did not improve bone mass. In contrast, high calcium suppressed serum 1,25-dihydroxyvitamin D levels and improved bone mass but increased the incidence of adenocarcinoma. Analysis of the VDR cistrome in RWPE1 prostate epithelial cells revealed vitamin D-mediated regulation of multiple cancer-relevant pathways. Our data support the hypothesis that the loss of vitamin D signaling accelerates the early stages of prostate carcinogenesis, and our results suggest that different dietary requirements may be needed to support prostate health or maximize bone mass. SIGNIFICANCE: This work shows that disrupting vitamin D signaling through diet or genetic deletion increases early prostate carcinogenesis through multiple pathways. Higher-diet vitamin D levels are needed for cancer than bone.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.