Merck
  • Isorhamnetin protects against hypoxia/reoxygenation-induced injure by attenuating apoptosis and oxidative stress in H9c2 cardiomyocytes.
Isorhamnetin protects against hypoxia/reoxygenation-induced injure by attenuating apoptosis and oxidative stress in H9c2 cardiomyocytes.
Gene (2018-05-08)
Ting-Ting Zhao, Tian-Lun Yang, Li Gong, Pei Wu
ABSTRACT
To unveil the possible protective role of isorhamnetin, an immediate 3'-O-methylated metabolite of quercetin, in cardiomyocyte under hypoxia/reoxygenation (H/R) condition and the underlying mechanisms involved, H9c2 cardiomyocytes were exposed to the vehicle or H/R for 6 h (2 h of hypoxia following by 4 h of reoxygenation) with isorhamnetin (0, 3, 6, 12, 25, 50 μM for 4 h prior to H/R exposure). Apoptosis was evaluated by TUNEL staining, flow cytometry analysis and western blot assay for cleaved caspase-3. Myocardial injure in vivo was determined by infarct size using TTC staining, histological damage using H&E staining and myocardial apoptosis. Here, we found that isorhamnetin dose-dependently protected H9c2 cardiomyocytes against H/R-induced injure, as evidenced by the reduction in lactate dehydrogenase (LDH) levels, increases in cell viability, superoxide dismutase (SOD) and catalase (CAT) activity, with the maximal effects at 25 μΜ. In addition, isorhamnetin treatment significantly inhibited apoptosis in H/R-induced H9c2 cardiomyocytes and ameliorated H/R-induced myocardial injure in vivo, concomitant with the upregulation of sirtuin 1 (SIRT1) expression. Mechanism studies demonstrated that isorhamnetin pretreatment remarkably abolished H/R-induced downregulation of Nuclear factor erythroid 2-related factor 2 (Nrf2) and heme oxygenase-1 (HO-1) expressions and upregulation of NADPH oxidase-2/4 (NOX-2/4) expressions in cardiomyocytes. However, SIRT1 inhibition (Sirtinol) not only inhibited isorhamnetin-induced Nrf2/HO-1 upregulation and NOX-2/4 downregulation, but also alleviated its anti-apoptotic effects. Taken together, these data indicate that isorhamnetin can exhibit positive effect on H/R-induced injure by attenuating apoptosis and oxidative stress in H9c2 cardiomyocytes, which is partly attributable to the upregulation of SIRT1 and Nrf2/HO-1-mediated antioxidant signaling pathway.
Sigma-Aldrich
L-(−)-Glucose, ≥99%