1,2–Dichloroethane solution
Synonym(s):
Ethylene chloride, Ethylene dichloride, 1,2–Dichloroethane solution, 1,2-Dichloroethane
Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing