Skip to Content
Merck
1-Butanol Pharmaceutical Secondary Standard; Certified Reference Material

1-Butanol

1-Butanol Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
n-Butanol, Butyl alcohol
Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Brand
Boiling Point (°C)
Color
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2