Skip to Content
Merck
Sodium nitrite analytical standard

Sodium nitrite

Sodium nitrite analytical standard
Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing