β-Carotene
Synonym(s):
β,β-Carotene, Provitamin A, β-Carotene
Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing