Butylated Hydroxytoluene
Synonym(s):
2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC, 3,5-Di-tert-butyl-4-hydroxytoluene, E321
Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing