Skip to Content
Merck
Arginine hydrochloride British Pharmacopoeia (BP) Reference Standard

Arginine Hydrochloride

Arginine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
L-Arginine monohydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride
Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique