L-Menthol
Synonym(s):
(−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol
Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique