Skip to Content
Merck
Nitric acid solution 9.5 % (w/w) in water

Nitric acid

Nitric acid solution 9.5 % (w/w) in water
Synonym(s):
Nitric acid solution
Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reaction Type
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2