Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trizma
®
hydrochloride solution
Synonym(s):
Trizma
®
hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride
Linear Formula:
NH
2
C(CH
2
OH)
3
· HCl
CAS Number:
1185-53-1
Molecular Weight:
157.60
EC Number:
214-684-5
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (2)
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SBR00051
Trizma
®
hydrochloride solution,
pH 8.5, DNase and Microbial DNA free, 10 mM
Expand
View Pricing
T6666
Trizma
®
hydrochloride solution,
pH 3.5-5.0 (0.5 M in H
2
O), BioXtra, ≥99.0% (titration)
Expand
View Pricing