Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Glutamine solution
Synonym(s):
L-Glutamine solution
CAS Number:
56-85-9
Products
Product Category
amino acids (2)
Brand
SAFC (1)
Sigma-Aldrich (2)
Color
Gene Alias
Physical Form
Product Line
Quality Segment
Optical Isomer
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G7513
L-Glutamine solution,
200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Expand
View Pricing
59202C
L-Glutamine solution,
29.23 mg/mL in saline, solution, suitable for cell culture
Expand
View Pricing
G2150
L-Glutamine solution,
200 mM, Hybri-Max
™
, sterile-filtered, suitable for hybridoma
Expand
View Pricing