Zinc chloride 0.1 M solution
Synonym(s):
Zinc chloride solution
Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core
Showing 1-4 of 4