Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Toluene solution
Synonym(s):
Toluene solution
CAS Number:
108-88-3
EC Number:
200-659-6
Products
Brand
Supelco (2)
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
40084
Toluene solution,
certified reference material, 5000 μg/mL in methanol
Expand
View Pricing
CRM48620
Toluene solution,
certified reference material,
Trace
CERT
®
, 200 μg/mL in methanol, ampule of 1 mL
Expand
View Pricing