Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zirconium(IV) oxide
Linear Formula:
ZrO
2
CAS Number:
1314-23-4
Molecular Weight:
123.22
Products
Brand
Sigma-Aldrich (12)
Sustainability
Feature
Formula Weight
pH Value
Physical Form
Chemical Composition
Quality Segment
Functional Group
Reagent Type
Core
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
643025
Zirconium(IV) oxide,
nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H
2
O
Expand
View Pricing
643122
Zirconium(IV) oxide,
nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H
2
O
Expand
View Pricing
916005
Zirconium(IV) oxide,
5 nm (core), Ethyl Acetate 50 wt. %, high surface coverage capping materials
Expand
View Pricing
915262
Zirconium(IV) oxide,
5 nm (core), methacrylate functionalized, 50 wt. % in propylene glycol methyl ether acetate (PGMEA), medium surface coverage capping materials
Expand
View Pricing
916269
Zirconium(IV) oxide,
5 nm (core), Propylene glycol methyl ether acetate (PGMEA) 50 wt. %, high surface coverage capping materials
Expand
View Pricing
915505
Zirconium(IV) oxide,
5 nm (core), methacrylate functionalized, 50 wt. % in propylene glycol methyl ether acetate (PGMEA)
Expand
View Pricing
916528
Zirconium(IV) oxide,
5 nm (core), methacrylate functionalized, Ethyl Acetate 50 wt. %, high surface coverage capping materials
Expand
View Pricing
915254
Zirconium(IV) oxide,
5 nm (core), acrylate functionalized, 50 wt. % in propylene glycol methyl ether acetate (PGMEA), medium surface coverage capping material
Expand
View Pricing
916501
Zirconium(IV) oxide,
5 nm (core), acrylate functionalized, Ethyl Acetate 50 wt. %, medium surface coverage capping materials
Expand
View Pricing
919985
Zirconium(IV) oxide,
5 nm (core), 50 wt. % in ethyl acetate, low surface coverage capping materials
Expand
View Pricing
915750
Zirconium(IV) oxide,
5 nm (core), methacrylate functionalized, 50 wt. % in ethyl acetate, medium surface coverage capping materials
Expand
View Pricing
915491
Zirconium(IV) oxide,
5 nm (core), 50 wt. % in propylene glycol methyl ether acetate (PGMEA), low surface coverage capping materials
Expand
View Pricing