Skip to Content
Merck
Glutaraldehyde solution 50% in H2O, suitable for photographic applications

Glutaraldehyde solution

Glutaraldehyde solution 50% in H2O, suitable for photographic applications
Synonym(s):
Glutaric dialdehyde solution, Pentane-1,5-dial, Glutaraldehyde solution
Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12

Products

Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Research Area
Quality Segment
Application
Organoleptic Class
Special Grade
Technique