Skip to Content
Merck
Sodium Chloride Tablets Each tablet yields 1 L of 0.9% sodium chloride.

Sodium Chloride Tablets

Sodium Chloride Tablets Each tablet yields 1 L of 0.9% sodium chloride.
Synonym(s):
Sodium chloride, Halite, NaCl
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44

Products

Brand
Formula Weight
Melting Point (Ā°C)
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing