Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ergocalciferol
Synonym(s):
Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D
2
Empirical Formula (Hill Notation):
C
28
H
44
O
CAS Number:
50-14-6
Molecular Weight:
396.65
EC Number:
200-014-9
Products
Product Category
vitamins (3)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
95220
Ergocalciferol,
≥98.0% (sum of enantiomers, HPLC)
Expand
View Pricing
E5750
Ergocalciferol,
40,000,000 USP units/g
Expand
View Pricing
E0900000
Ergocalciferol,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP788
Ergocalciferol,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing