Skip to Content
Merck
DL-Homocysteine analytical standard

DL-Homocysteine

DL-Homocysteine analytical standard
Synonym(s):
2-Amino-4-mercaptobutyric acid
Linear Formula:
HSCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
135.18
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Analyte Suitability
Quality Segment
Application
Amino Acid
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing