Skip to Content
Merck
1,2-Propanediol

Propylene glycol

1,2-Propanediol
Synonym(s):
1,2-Propanediol, Propylene glycol
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique