Kanamycin sulfate from Streptomyces kanamyceticus
Synonym(s):
Kanamycin A, Kanamycin sulfate salt, Bekanamycin sulfate salt, Kanamycin A sulfate salt, Kanamycin sulfate from Streptomyces kanamyceticus
Empirical Formula (Hill Notation):
C18H37N5O10 · xH2SO4
CAS Number:
Molecular Weight:
483.51 (free base basis)
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing