Skip to Content
Merck
(−)-Epigallocatechin gallate ≥80% (HPLC), from green tea

(−)-Epigallocatechin gallate

(−)-Epigallocatechin gallate ≥80% (HPLC), from green tea
Synonym(s):
(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG
Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Purity
Research Area
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing