Skip to Content
Merck
Sodium pyrophosphate tetrabasic decahydrate BioUltra, ≥99.5% (T)

Sodium pyrophosphate decahydrate

Sodium pyrophosphate tetrabasic decahydrate BioUltra, ≥99.5% (T)
Synonym(s):
Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate, tetra-Sodium diphosphate decahydrate, Pyrophosphate inorganic, Sodium diphosphate tetrabasic
Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade