Sodium periodate
Synonym(s):
Sodium (meta)periodate, Sodium periodate, Sodium metaperiodate
Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Special Grade