Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DCC
Synonym(s):
N
,
N
′-Dicyclohexylcarbodiimide, N,N′-Dicyclohexylcarbodiimide
Linear Formula:
C
6
H
11
N=C=NC
6
H
11
CAS Number:
538-75-0
Molecular Weight:
206.33
EC Number:
208-704-1
Products
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Puriss (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D80002
DCC,
99%
Expand
View Pricing
8.02954
DCC,
for synthesis
Expand
View Pricing
36650
DCC,
puriss., ≥99.0% (GC)
Expand
View Pricing