Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-(Methylamino)phenol hemisulfate salt
Synonym(s):
Metol, 4-(Methylamino)phenol hemisulfate salt
Linear Formula:
C
7
H
9
NO · 1/2H
2
SO
4
CAS Number:
55-55-0
Molecular Weight:
172.19
EC Number:
200-237-1
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
ACS reagent (1)
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
320013
4-(Methylamino)phenol hemisulfate salt,
ACS reagent, 99%
Expand
View Pricing
1.07299
4-(Methylamino)phenol hemisulfate salt,
Technipur
®
Expand
View Pricing
69750
4-(Methylamino)phenol hemisulfate salt,
purum, for photographic purposes, ≥98.0% (HPLC)
Expand
View Pricing
15501
4-(Methylamino)phenol hemisulfate salt,
≥98% (HPLC)
Expand
View Pricing