Diethyl pyrocarbonate
Synonym(s):
DEP, DEPC, Diethyl dicarbonate, Diethyl oxydiformate, Ethoxyformic acid anhydride
Linear Formula:
O(COOC2H5)2
CAS Number:
Molecular Weight:
162.14
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing