Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium thiosulfate
Synonym(s):
Sodium thiosulfate
Linear Formula:
Na
2
S
2
O
3
CAS Number:
7772-98-7
Molecular Weight:
158.11
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Anhydrous (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
217263
Sodium thiosulfate,
ReagentPlus
®
, 99%
Expand
View Pricing
72049
Sodium thiosulfate,
purum p.a., anhydrous, ≥98.0% (RT)
Expand
View Pricing
1.06512
Sodium thiosulfate,
anhydrous EMPLURA
®
Expand
View Pricing
563188
Sodium thiosulfate,
≥99.99% trace metals basis
Expand
View Pricing
909173
Sodium thiosulfate,
free-flowing, Redi-Dri
™
,
ReagentPlus
®
, 99%
Expand
View Pricing