Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium iron(III) citrate
Synonym(s):
Ammonium ferric citrate, Ferric ammonium citrate, FRASER Listeria Ammonium iron(III) Supplement, Ammonium iron(III) citrate, Amonium Iron(III) Citrate Supplement
Linear Formula:
C
6
H
8
O
7
·xFe
3
+
· yNH
3
CAS Number:
1185-57-5
EC Number:
214-686-6
Products
Product Category
salts (3)
media supplements (2)
Brand
Millipore (2)
SAFC (2)
Sigma-Aldrich (1)
Biological Source
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
F5879
Ammonium iron(III) citrate,
reagent grade, powder
Expand
View Pricing
1.00092
Ammonium iron(III) citrate,
for
Listeria
spp., pkg of 10 vials, for the preparation of Half FRASER (Demi FRASER) Broth (Base) or FRASER Broth (Base)
Expand
View Pricing
RES20400-A7
Ammonium iron(III) citrate,
Expand
View Pricing
1.03762
Ammonium iron(III) citrate,
about 18% Fe
Expand
View Pricing
90836
Ammonium iron(III) citrate,
suitable for microbiology
Expand
View Pricing