Skip to Content
Merck
All Photos(1)

Key Documents

171263-M

Millipore

AMPK Signaling Activator IX, F17

The AMPK Signaling Agonist, F17, also referenced under CAS 280112-24-5, controls the biological activity of AMPK. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

AMPK Signaling Activator IX, F17, 4-hydroxy-1-isobutyl-N-(5-methylthiazol-2-yl)-2-oxo-1,2-dihydroquinoline-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19N3O3S
CAS Number:
Molecular Weight:
357.43
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL, clear, colorless

storage temp.

2-8°C

InChI

1S/C18H19N3O3S/c1-10(2)9-21-13-7-5-4-6-12(13)15(22)14(17(21)24)16(23)20-18-19-8-11(3)25-18/h4-8,10,22H,9H2,1-3H3,(H,19,20,23)

InChI key

QSZAFLFWFQTCJK-UHFFFAOYSA-N

General description

A cell-permeable 4-hydroxy-2-oxo quinoline carboxamide that acts as an agonist of AMPK signaling. It is shown to reduce fat storage in C. elegans (EC50 ~ 0.1 µM) and reduce the expression level of fat-7 (a C. elegans ortholog of mammalian stearoyl-CoA desaturase-1), through aak-1 (an ortholog for the AMPK-α1 catalytic subunit) and K08F8.2 (a transcription factor) dependent mechanisms. In addition, it markedly reduces the number of lipid droplets in HepG2 human hepatocarcinoma cells at 25 µM, and elicits the phosphorylation and inactivation of acetyl coenzyme A carboxylase (ACC, the enzyme that catalyzes the rate-limiting step in de novo fatty acid synthesis). F17-induced ACC phosphorylation can be abrogated by simultaneous treatment with an AMPK inhibitor compound C (Cat. No. 171260).

Warning

Toxicity: Standard Handling (A)

Preparation Note

Warming to 55-60°C is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lemieux, G.A., et al. 2011. Nat. Chem. Biol.7, 206.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service