- Coexistence of t(15;17) and t(15;16;17) detected by fluorescence
Coexistence of t(15;17) and t(15;16;17) detected by fluorescence
Oncology letters (2014-08-15)
Rui Zhang, Young-Mi Kim, Xianfu Wang, Yan Li, Hui Pang, Ji-Yun Lee, Shibo Li
PMID25120648
RÉSUMÉ
Acute promyelocytic leukemia (APL) is characterized by the t(15;17)(q22;q21), which results in the fusion of the promyelocytic leukemia (