Merck
Deciphering anti-MOG IgG antibodies: Clinical and radiological spectrum, and comparison of antibody detection assays.
Journal of the neurological sciences (2020-01-19)
John S Tzartos, Katerina Karagiorgou, Dimitrios Tzanetakos, Marianthi Breza, Maria Eleftheria Evangelopoulos, Sygkliti-Henrietta Pelidou, Christos Bakirtzis, Ioannis Nikolaidis, Georgios Koutsis, Konstantinos Notas, Elisabeth Chroni, Ioannis Markakis, Nikolaos C Grigoriadis, Maria Anagnostouli, Anastasios Orologas, Dimitrios Parisis, Theodoros Karapanayiotides, Dimitra Papadimitriou, Vasiliki Kostadima, John Elloul, Iosif Xidakis, Thomas Maris, Paraskevi Zisimopoulou, Socrates Tzartos, Costas Kilidireas
RÉSUMÉ
IgG antibodies to myelin oligodendrocyte glycoprotein (MOG) detected by cell based assays (CBA) have been identified in a constantly expanding spectrum of CNS demyelinating disorders. However, a universally accepted CBA has not been adopted yet. We aimed to analyze the clinical and radiological features of patients with anti-MOG IgG1-antibodies detected with a live-cell CBA and to compare the three most popular MOG-CBAs. We screened sera from 1300 Greek patients (including 426 patients referred by our 8 clinics) suspected for anti-MOG syndrome, and 120 controls with the live-cell MOG-CBA for IgG1-antibodies. 41 patients, versus 0 controls were seropositive. Clinical, serological and radiological data were available and analyzed for the 21 seropositive patients out of the 426 patients of our clinics. Their phenotypes were: 8 optic neuritis, 3 myelitis, 3 neuromyelitis optica, 2 encephalomyelitis, 2 autoimmune encephalitis and 3 atypical MS. We then retested all sera of our 426 patients with the other two most popular MOG-CBAs for total IgG (a live-cell and a commercial fixed-cell CBAs). Seven IgG1-seropositive patients were seronegative for one or both IgG-CBAs. Yet, all 21 patients had clinical and radiological findings previously described in MOG-antibody associated demyelination disease supporting the high specificity of the IgG1-CBA. In addition, all IgG1-CBA-negative sera were also negative by the IgG-CBAs. Also, all controls were negative by all three assays, except one serum found positive by the live IgG-CBA. Overall, our findings support the wide spectrum of anti-MOG associated demyelinating disorders and the superiority of the MOG-IgG1 CBA over other MOG-CBAs.