Principaux documents
SYMBISL01WW
Millipore
Steritest Symbio ISL Pump Kit, 2 media
About This Item
Code UNSPSC :
23151818
eCl@ss :
32014001
Nomenclature NACRES :
JA.12
Matériaux
316L stainless steel
Niveau de qualité
Gamme de produits
Steritest®
Poids
17.6 kg (pump body) with standard accessories:19.2 kg
Description générale
The Steritest Symbio ISL Pump is optimized for more ergonomic sterility testing inside isolators. Its table-integrated design offers more working space and loading volume in isolators.
Application
The Steritest Symbio ISL Pump, designed to fit your testing environment, Sterility Testing.
Informations légales
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Clause de non-responsabilité
Sterility testing USP 71, EP2.6.1 , JP 4.06