Merck
  • Human cactin interacts with DHX8 and SRRM2 to assure efficient pre-mRNA splicing and sister chromatid cohesion.
Human cactin interacts with DHX8 and SRRM2 to assure efficient pre-mRNA splicing and sister chromatid cohesion.
Journal of cell science (2017-01-08)
Isabella M Y Zanini, Charlotte Soneson, Luca E Lorenzi, Claus M Azzalin
ABSTRACT
Cactins constitute a family of eukaryotic proteins broadly conserved from yeast to human and required for fundamental processes such as cell proliferation, genome stability maintenance, organismal development and immune response. Cactin proteins have been found to associate with the spliceosome in several model organisms, nevertheless their molecular functions await elucidation. Here we show that depletion of human cactin leads to premature sister chromatid separation, genome instability and cell proliferation arrest. Moreover, cactin is essential for efficient splicing of thousands of pre-mRNAs, and incomplete splicing of the pre-mRNA of sororin (also known as CDCA5), a cohesin-associated factor, is largely responsible for the aberrant chromatid separation in cactin-depleted cells. Lastly, cactin physically and functionally interacts with the spliceosome-associated factors DHX8 and SRRM2. We propose that cellular complexes comprising cactin, DHX8 and SRRM2 sustain precise chromosome segregation, genome stability and cell proliferation by allowing faithful splicing of specific pre-mRNAs. Our data point to novel pathways of gene expression regulation dependent on cactin, and provide an explanation for the pleiotropic dysfunctions deriving from cactin inactivation in distant eukaryotes.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-TRF2 Antibody, clone 4A794, clone 4A794, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse