Merck
  • HPW-RX40 prevents human platelet activation by attenuating cell surface protein disulfide isomerases.
HPW-RX40 prevents human platelet activation by attenuating cell surface protein disulfide isomerases.
Redox biology (2017-06-11)
Po-Hsiung Kung, Pei-Wen Hsieh, Ying-Ting Lin, Jia-Hau Lee, I-Hua Chen, Chin-Chung Wu
ABSTRACT
Protein disulfide isomerase (PDI) present at platelet surfaces has been considered to play an important role in the conformational change and activation of the integrin glycoprotein IIb/IIIa (GPIIb/IIIa) and thus enhances platelet aggregation. Growing evidences indicated that platelet surface PDI may serve as a potential target for developing of a new class of antithrombotic agents. In the present study, we investigated the effects of HPW-RX40, a chemical derivative of β-nitrostyrene, on platelet activation and PDI activity. HPW-RX40 inhibited platelet aggregation, GPIIb/IIIa activation, and P-selectin expression in human platelets. Moreover, HPW-RX40 reduced thrombus formation in human whole blood under flow conditions, and protects mice from FeCl
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film