Merck
  • CaMKIIδ meditates phenylephrine induced cardiomyocyte hypertrophy through store-operated Ca
CaMKIIδ meditates phenylephrine induced cardiomyocyte hypertrophy through store-operated Ca
Cardiovascular pathology : the official journal of the Society for Cardiovascular Pathology (2016-12-13)
Yawei Ji, Xin Guo, Zhe Zhang, Zhuyun Huang, Jianghua Zhu, Qing-Hui Chen, Le Gui
ABSTRACT
Evidence suggests that store-operated Ca2+ entry (SOCE) is involved in the hypertrophy of cardiomyocytes. The signaling mechanisms of SOCE contributing to cardiac hypertrophy following phenylephrine (PE) stimulation are not fully understood. Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human MYH1
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film