Merck
  • Preventive and therapeutic effects of gene therapy using silica nanoparticles-binding of GM-CSF gene on white blood cell production in dogs with leukopenia.
Preventive and therapeutic effects of gene therapy using silica nanoparticles-binding of GM-CSF gene on white blood cell production in dogs with leukopenia.
Experimental hematology (2008-06-14)
Eun Wha Choi, Hye Cheong Koo, Il Seob Shin, Young Jin Chae, Jong Hwa Lee, Sei Myoung Han, Seung Jun Lee, Dong Ha Bhang, Yong Ho Park, Chang Woo Lee, Hwa Young Youn
ABSTRACT
Our previous study has shown that granulocyte-macrophage colony-stimulating factor (GM-CSF) gene/silica nanoparticles have a leukocytosis effect in normal dogs. Therefore, this study was conducted to determine whether treatment of canine GM-CSF gene/silica nanoparticles has preventive or therapeutic effects in dogs with leukopenia. To induce leukopenia, vinblastine was administered intravenously at a dose of 2 mg/m(2) of body surface area on day 0. Then 7.5 microg GM-CSF/nanoparticles (1:100, w/w) were administered intravenously to each of four dogs in the prevention group on day 2 and an equivalent amount of GM-CSF/nanoparticles was administered to the post-nadir group on day 4 (other groups were administered phosphate-buffered saline intravenously). Therapeutic GM-CSF gene was expressed in peripheral blood mononuclear cells for 10 days and both the prevention and post-nadir groups showed significant increases in white blood cell counts when compared with the control group, as confirmed by complete blood count, differential count, and flow cytometry. GM-CSF/nanoparticles can be useful for correction of acute leukopenia, such as chemotherapy-induced myelosuppression, without developing neutralizing antibodies.