Merck
  • Mevastatin ameliorates sphingosine 1-phosphate-induced COX-2/PGE2-dependent cell migration via FoxO1 and CREB phosphorylation and translocation.
Mevastatin ameliorates sphingosine 1-phosphate-induced COX-2/PGE2-dependent cell migration via FoxO1 and CREB phosphorylation and translocation.
British journal of pharmacology (2015-09-12)
Chih-Kai Hsu, Chih-Chung Lin, Li-Der Hsiao, Chuen-Mao Yang
ABSTRACT
Sphingosine 1-phosphate (S1P), an important inflammatory mediator, has been shown to regulate COX-2 production and promote various cellular responses such as cell migration. Mevastatin, an inhibitor of 3-hydroxy-3-methylglutaryl-CoA reductase (HMG-CoA), effectively inhibits inflammatory responses. However, the mechanisms underlying S1P-evoked COX-2-dependent cell migration, which is modulated by mevastatin in human tracheal smooth muscle cells (HTSMCs) remain unclear. The expression of COX-2 was determined by Western blotting, real time-PCR and promoter analyses. The signalling molecules were investigated by pretreatment with respective pharmacological inhibitors or transfection with siRNAs. The interaction between COX-2 promoter and transcription factors was determined by chromatin immunoprecipitation assay. Finally, the effect of mevastatin on HTSMC migration and leukocyte counts in BAL fluid and COX-2 expression induced by S1P was determined by a cell migration assay, cell counting and Western blot. S1P stimulated mTOR activation through the Nox2/ROS and PI3K/Akt pathways, which can further stimulate FoxO1 phosphorylation and translocation to the cytosol. We also found that S1P induced CREB activation and translocation via an mTOR-independent signalling pathway. Finally, we showed that pretreatment with mevastatin markedly reduced S1P-induced cell migration and COX-2/PGE2 production via a PPARγ-dependent signalling pathway. Mevastatin attenuates the S1P-induced increased expression of COX-2 and cell migration via the regulation of FoxO1 and CREB phosphorylation and translocation by PPARγ in HTSMCs. Mevastatin could be beneficial for prevention of airway inflammation in the future.
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein, ≥90% (HPLC)
Sigma-Aldrich
GW9662, >98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder