Merck
  • CREB-induced inflammation is important for malignant mesothelioma growth.
CREB-induced inflammation is important for malignant mesothelioma growth.
The American journal of pathology (2014-08-12)
Catherine M Westbom, Anurag Shukla, Maximilian B MacPherson, Elizabeth C Yasewicz, Jill M Miller, Stacie L Beuschel, Chad Steele, Harvey I Pass, Pamela M Vacek, Arti Shukla
ABSTRACT
Malignant mesothelioma (MM) is an aggressive tumor with no treatment regimen. Previously we have demonstrated that cyclic AMP response element binding protein (CREB) is constitutively activated in MM tumor cells and tissues and plays an important role in MM pathogenesis. To understand the role of CREB in MM tumor growth, we generated CREB-inhibited MM cell lines and performed in vitro and in vivo experiments. In vitro experiments demonstrated that CREB inhibition results in significant attenuation of proliferation and drug resistance of MM cells. CREB-silenced MM cells were then injected into severe combined immunodeficiency mice, and tumor growth in s.c. and i.p. models of MM was followed. We observed significant inhibition in MM tumor growth in both s.c. and i.p. models and the presence of a chemotherapeutic drug, doxorubicin, further inhibited MM tumor growth in the i.p. model. Peritoneal lavage fluids from CREB-inhibited tumor-bearing mice showed a significantly reduced total cell number, differential cell counts, and pro-inflammatory cytokines and chemokines (IL-6, IL-8, regulated on activation normal T cell expressed and secreted, monocyte chemotactic protein-1, and vascular endothelial growth factor). In vitro studies showed that asbestos-induced inflammasome/inflammation activation in mesothelial cells was CREB dependent, further supporting the role of CREB in inflammation-induced MM pathogenesis. In conclusion, our data demonstrate the involvement of CREB in the regulation of MM pathogenesis by regulation of inflammation.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride