Merck
  • SCIMP, a transmembrane adaptor protein involved in major histocompatibility complex class II signaling.
SCIMP, a transmembrane adaptor protein involved in major histocompatibility complex class II signaling.
Molecular and cellular biology (2011-09-21)
Peter Draber, Ivana Vonkova, Ondrej Stepanek, Matous Hrdinka, Marketa Kucova, Tereza Skopcova, Pavel Otahal, Pavla Angelisova, Vaclav Horejsi, Mandy Yeung, Arthur Weiss, Tomas Brdicka
ABSTRACT
Formation of the immunological synapse between an antigen-presenting cell (APC) and a T cell leads to signal generation in both cells involved. In T cells, the lipid raft-associated transmembrane adaptor protein LAT plays a central role. Its phosphorylation is a crucial step in signal propagation, including the calcium response and mitogen-activated protein kinase activation, and largely depends on its association with the SLP76 adaptor protein. Here we report the discovery of a new palmitoylated transmembrane adaptor protein, termed SCIMP. SCIMP is expressed in B cells and other professional APCs and is localized in the immunological synapse due to its association with tetraspanin-enriched microdomains. In B cells, it is constitutively associated with Lyn kinase and becomes tyrosine phosphorylated after major histocompatibility complex type II (MHC-II) stimulation. When phosphorylated, SCIMP binds to the SLP65 adaptor protein and also to the inhibitory kinase Csk. While the association with SLP65 initiates the downstream signaling cascades, Csk binding functions as a negative regulatory loop. The results suggest that SCIMP is involved in signal transduction after MHC-II stimulation and therefore serves as a regulator of antigen presentation and other APC functions.