Merck
  • MicroRNAs are critical regulators of tuberous sclerosis complex and mTORC1 activity in the size control of the Xenopus kidney.
MicroRNAs are critical regulators of tuberous sclerosis complex and mTORC1 activity in the size control of the Xenopus kidney.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-16)
Daniel Romaker, Vikash Kumar, Débora M Cerqueira, Ryan M Cox, Oliver Wessely
ABSTRACT
MicroRNAs (miRNAs) are major posttranscriptional regulators of a wide variety of biological processes. However, redundancy among most miRNAs has made it difficult to identify their in vivo functions. We previously demonstrated that global inhibition of miRNA biogenesis in Xenopus resulted in a dramatically smaller pronephric kidney. This suggested that microRNAs play a pivotal role in organ size control. Here we now provide a detailed mechanistic explanation for this phenotype. We identified that the activation of the mechanistic target of rapamycin complex 1 (mTORC1) by Insulin and insulin-like growth factor (Igf) 2 is an important regulator in kidney growth, which in turn is modulated by microRNAs. Molecular analyses demonstrate that microRNAs set a threshold for mTORC1 signaling by down-regulating one of its core negative regulators, tuberous sclerosis 1 (Tsc1). Most importantly, this rheostat can be reprogrammed experimentally. Whereas knockdown of miRNAs causes growth arrest, concomitant knockdown of Tsc1 restores mTORC1 activity and proximal tubular size. Together, these data establish a previously unidentified in vivo paradigm for the importance of posttranscriptional regulation in organ size control.
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Sigma-Aldrich
Anti-phospho-ERK1 (pThr202/pTyr204) and ERK2 (pThr185/pTyr187) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)