Merck
Switchable viscosity triggered by CO2 using smart worm-like micelles.
Chemical communications (Cambridge, England) (2013-02-26)
Xin Su, Michael F Cunningham, Philip G Jessop
ABSTRACT
We report two CO2-responsive aqueous solutions having switchable viscosity. The solution of sodium octadecyl sulfate and 2-(dimethylamino)ethanol forms viscoelastic worm-like micelles in the presence of CO2 but not in its absence at 60 °C. Another solution of sodium stearate and sodium nitrate has high viscosity only if CO2 is absent.
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%