Merck
  • A divergent approach to 3-piperidinols: a concise syntheses of (+)-swainsonine and access to the 1-substituted quinolizidine skeleton.
A divergent approach to 3-piperidinols: a concise syntheses of (+)-swainsonine and access to the 1-substituted quinolizidine skeleton.
The Journal of organic chemistry (2012-08-16)
Glenn Archibald, Chih-Pei Lin, Peter Boyd, David Barker, Vittorio Caprio
ABSTRACT
Nucleophilic addition of Grignard reagents and organolithium species to a 3-silyloxy-3,4,5,6-tetrahydropyridine N-oxide provides trans-2,3-disubstituted N-hydroxypiperidines exclusively. The application of this methodology to the preparation of a diversity of useful trans-2-substituted-3-hydroxypiperidines, a concise synthesis of (+)-swainsonine, and an enantiopure 1-substituted quinolizidine of utility in target-directed synthesis is reported.
Sigma-Aldrich
Swainsonine, synthetic
Sigma-Aldrich
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)