Merck
  • Anticancer efficacy of deguelin in human prostate cancer cells targeting glycogen synthase kinase-3 β/β-catenin pathway.
Anticancer efficacy of deguelin in human prostate cancer cells targeting glycogen synthase kinase-3 β/β-catenin pathway.
International journal of cancer (2011-04-08)
Vijayalakshmi Thamilselvan, Mani Menon, Sivagnanam Thamilselvan
ABSTRACT
Activation of survival pathways has been associated with chemoresistance and progression of androgen independence which places a major obstacle to successful treatment of metastatic prostate cancer. Deguelin, a rotenoid isolated from Mundulea sericea, has an anticancer effect against several types of cancers; however, the mechanism of its antitumor effects on prostate cancer is not well understood. The aim of our study was to elucidate the effect of deguelin on the growth of prostate cancer cells and its putative mechanism of action. Deguelin decreased the viability of both androgen-dependent and -independent prostate cancer cells but not normal prostate epithelial cells. Downregulation of phosphorylated Akt and GSK-3β by deguelin promoted proteosomal degradation of β-catenin that resulted in decreased nuclear accumulation and inhibited transactivation of β-catenin-responsive genes. Deguelin-induced downregulation of proliferative (cyclin D1 and c-myc) and antiapoptotic proteins (Mcl-1, Bcl-xL and survivin) in prostate cancer cells culminated in the induction of apoptosis, inhibition of DNA synthesis and cell growth, altered membrane integrity, marked reduction of invasiveness, inhibition of anchorage-dependent and -independent colony formation. Our data demonstrated for the first time that deguelin inhibits the growth and survival of human androgen-independent prostate cancer cells, and its anticancer and antimetastatic activity occurs, at least in part through downregulating GSK-3β/β-catenin signaling pathway and antiapoptotic survival proteins. Taken together our study indicates that deguelin may have translational potential as therapeutic agent for advanced or metastatic prostate cancer.
Sigma-Aldrich
(−)-Deguelin, >98% (HPLC), powder