Merck
  • Stereoselective synthesis of tetrasubstituted (Z)-alkenes from aryl alkyl ketones utilizing the Horner-Wadsworth-Emmons reaction.
Stereoselective synthesis of tetrasubstituted (Z)-alkenes from aryl alkyl ketones utilizing the Horner-Wadsworth-Emmons reaction.
Chemical & pharmaceutical bulletin (2002-09-19)
Shigeki Sano, Tomoka Takehisa, Shiho Ogawa, Kenji Yokoyama, Yoshimitsu Nagao
ABSTRACT
Tetrasubstituted (Z)-alkenes were readily prepared through the Horner-Wadsworth-Emmons reactions of methyl 2-[bis(2,2,2-trifluoroethyl)phosphono]propionate with aryl alkyl ketones by employing Sn(OSO(2)CF(3))(2) and N-ethylpiperidine.
Sigma-Aldrich
1-Ethylpiperidine, 99%