Merck
Neurturin, a relative of glial-cell-line-derived neurotrophic factor.
Nature (1996-12-05)
P T Kotzbauer, P A Lampe, R O Heuckeroth, J P Golden, D J Creedon, E M Johnson, J Milbrandt
ABSTRACT
The normal development of the vertebrate nervous system entails the death of 30-70% of the neurons originally generated in most neuronal populations. This naturally occurring cell death is regulated by specific neurotrophic factors that promote neuronal survival and which are produced in limiting quantities by target cells, glial cells and neurons. These factors are also of potential utility as therapeutic agents for neurodegenerative diseases. Here we describe the purification and cloning of a new neurotrophic factor, identified on the basis of its ability to support the survival of sympathetic neurons in culture. This factor, neurturin, is structurally related to glial-cell-line-derived neurotrophic factor (GDNF). These factors can each activate the MAP kinase signalling pathway in cultured sympathetic neurons and support the survival of sympathetic neurons, as well as of sensory neurons of the nodose and dorsal root ganglia. Thus, neurturin and GDNF together now define a new family of neurotrophic factors.
Sigma-Aldrich
Neurturin human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture