Merck
  • Involvement of intracellular signaling in the IL-1β inhibitory effect on fructose intestinal absorption.
Involvement of intracellular signaling in the IL-1β inhibitory effect on fructose intestinal absorption.
Journal of cellular physiology (2014-09-13)
María J Rodríguez-Yoldi, Sonia Gascón, Cristina Barranquero, Alberto García-Barrios, Jesús Osada
ABSTRACT
A variety of bacteria and their excreted/secreted products having direct effects on epithelial ion transport and permeability and the release of cytokines during bacterial infection may impact directly on epithelial function. Interleukin-1β (IL-1β) is a pleiotropic cytokine that affects the intestinal absorption of nutrients. The aim of this work was to study the intracellular signaling pathways involved in the inhibitory effect of IL-1β on D-fructose intestinal transport in rabbit jejunum and Caco-2 cells. The results show that the cytokine inhibitory effect was completely reversed in presence of proteasome or PKC selective inhibitors in IL-1β treated rabbits. In addition, the activation of PI3K abolished the IL-1β effect. Likewise, these results were confirmed in Caco-2 cells. In addition, p-PI3K expression was increased by IL-1β-treatment whereas the expression of p-PKCα was not significantly affected. In summary, the results suggest that IL-1β could regulate the activation of pPKCα 73, pPI3K 55, and NF-kB proteins. These events could exert an inhibitory effect on fructose intestinal absorption by a modification of GLUT5 insertion to brush-border membrane and/or the functional transporter activity. This effect is independent of hormonal milieu and nervous stimuli.
Sigma-Aldrich
Goat Anti-Mouse light chain Antibody, HRP conjugate, Chemicon®, from goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution