Merck
  • Expression profiles of the internal jugular and saphenous veins: Focus on hemostasis genes.
Expression profiles of the internal jugular and saphenous veins: Focus on hemostasis genes.
Thrombosis research (2020-05-22)
Nicole Ziliotto, Silvia Meneghetti, Erica Menegatti, Marcello Baroni, Barbara Lunghi, Fabrizio Salvi, Manuela Ferracin, Alessio Branchini, Donato Gemmati, Francesco Mascoli, Paolo Zamboni, Francesco Bernardi, Giovanna Marchetti
ABSTRACT
Venous bed specificity could contribute to differential vulnerability to thrombus formation, and is potentially reflected in mRNA profiles. Microarray-based transcriptome analysis in wall and valve specimens from internal jugular (IJV) and saphenous (SV) veins collected during IJV surgical reconstruction in patients with impaired brain outflow. Multiplex antigenic assay in paired jugular and peripheral plasma samples. Most of the top differentially expressed transcripts have been previously associated with both vascular and neurological disorders. Large expression differences of HOX genes, organ patterning regulators, pinpointed the vein positional identity. The "complement and coagulation cascade" emerged among enriched pathways. In IJV, upregulation of genes for coagulation inhibitors (TFPI, PROS1), activated protein C pathway receptors (THBD, PROCR), fibrinolysis activators (PLAT, PLAUR), and downregulation of the fibrinolysis inhibitor (SERPINE1) and of contact/amplification pathway genes (F11, F12), would be compatible with a thromboprotective profile in respect to SV. Further, in SV valve the prothrombinase complex genes (F5, F2) were up-regulated and the VWF showed the highest expression. Differential expression of several VWF regulators (ABO, ST3GAL4, SCARA5, CLEC4M) was also observed. Among other differentially expressed hemostasis-related genes, heparanase (HPSE)/heparanase inhibitor (HPSE2) were up-/down-regulated in IJV, which might support procoagulant features and disease conditions. The jugular plasma levels of several proteins, encoded by differentially expressed genes, were lower and highly correlated with peripheral levels. The IJV and SV rely on differential expression of many hemostasis and hemostasis-related genes to balance local hemostasis, potentially related to differences in vulnerability to thrombosis.
Product Description
Millipore
MILLIPLEX® Human Angiogenesis/Growth Factor Magnetic Bead Panel - Cancer Multiplex Assay, Angiogenesie Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple angiogenic biomarkers in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 3 - Neuroscience Multiplex Assay, The analytes available for this multiplex kit are: BDNF, Cathepsin D, MPO, NCAM, PAI-1 (total), PDGF-AA, PDGF-AB/BB, RANTES, sICAM-1, sVCAM-1.