Merck
  • A Double-Sandwich ELISA for Identification of Monoclonal Antibodies Suitable for Sandwich Immunoassays.
A Double-Sandwich ELISA for Identification of Monoclonal Antibodies Suitable for Sandwich Immunoassays.
Methods in molecular biology (Clifton, N.J.) (2015-07-15)
Larry H Stanker, Robert M Hnasko
ABSTRACT
The sandwich immunoassay (sELISA) is an invaluable technique for concentrating, detecting, and quantifying target antigens. The two critical components required are a capture antibody and a detection antibody, each binding a different epitope on the target antigen. The specific antibodies incorporated into the test define most of the performance parameters of any subsequent immunoassay regardless of the assay format: traditional ELISA, lateral-flow immunoassay, various bead-based assays, antibody-based biosensors, or the reporting label. Here we describe an approach for identifying monoclonal antibodies (mAbs) suitable for use as capture antibodies and detector antibodies in a sELISA targeting bacterial protein toxins. The approach was designed for early identification of monoclonal antibodies (mAbs), in the initial hybridoma screen.
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution