Merck
  • Knockdown of long noncoding RNA dreh facilitates cell surface GLUT4 expression and glucose uptake through the involvement of vimentin in 3T3-L1 adipocytes.
Knockdown of long noncoding RNA dreh facilitates cell surface GLUT4 expression and glucose uptake through the involvement of vimentin in 3T3-L1 adipocytes.
Gene (2020-02-06)
Nobuhiko Takahashi, Atsushi P Kimura, Kazumasa Ohmura, Sumiyoshi Naito, Mika Yoshida, Masahiro Ieko
ABSTRACT
Glucose uptake in adipocytes is crucial for regulating systemic metabolism. Long noncoding RNAs (lncRNAs), defined as being transcripts with lengths exceeding 200 nucleotides that are not translated, are recently identified regulators of cellular functions. Previously, we have shown that an lncRNA, "down-regulated expression by hepatitis B virus X" (dreh), is involved in glucose transport in skeletal muscle cells. Here, we aimed to examine the involvement of dreh in glucose transport in 3T3-L1 adipocytes. Expression analysis showed that dreh was expressed in 3T3-L1 fibroblasts and adipocytes. Knockdown of dreh expression using its specific siRNAs lowered the glucose concentration of the medium and facilitated [3H]-2-deoxyglucose transport in adipocytes. Additionally, dreh silencing enhanced the protein expression of glucose transporter (GLUT4) in the plasma membrane of adipocytes. Treatment with siRNA against vimentin attenuated the glucose-lowering effect of dreh depletion. These results suggest that the repression of dreh facilitates glucose transport via increased GLUT4 expression in the plasma membrane through the involvement of vimentin in 3T3-L1 adipocytes. In conclusion, dreh is the first observed lncRNA that regulates glucose transport in adipocytes and could serve as a novel therapeutic target for diabetes by modulating adipocyte function. Considering the new function of dreh, we propose that dreh be renamed "down-regulated expression-related hexose/glucose transport enhancer."
Sigma-Aldrich
L-(−)-Glucose, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glucose (GO) Assay Kit, sufficient for 20 assays